CAS RN 1229652-21-4
CAS RN 1229652-21-4
IUPAC Name:
(3-{[4-({3-[(4-fluorophenyl)methyl]-2,4-dioxo-1,3-thiazolidin-5-ylidene}methyl)phenoxy]methyl}phenyl)boronic acid
Synonyms:
3-[4-({3-[(4-fluorophenyl)methyl]-2,4-dioxo-1,3-thiazolidin-5-ylidene}methyl)phenoxymethyl]phenylboronic acid
Molecular Weight (g/mol):
463.29
Molecular Formula:
C24H19BFNO5S
InChi Key:
VTNKMYWFWQTEHE-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(COC2=CC=C(C=C3SC(=O)N(CC4=CC=C(F)C=C4)C3=O)C=C2)=CC=C1
1
–
2
of
2
results
1
Tocris Bioscience™ HA 130

CAS: 1229652-21-4 Molecular Formula: C24H19BFNO5S Molecular Weight (g/mol): 463.286
2
Medchem Express HA130, MedChemExpress
MedChemExpress HA130 is a selective autotaxin (ATX) inhibitor with an IC50 of 28 nM.
