CAS RN 123-73-9
CAS RN 123-73-9
IUPAC Name:
(2E)-but-2-enal
Synonyms:
crotonaldehyde 1-formylpropene crotylaldehyde methylpropenal β-methylacrolein aldehyde crotonique propylene aldehyde
Molecular Weight (g/mol):
70.09
Molecular Formula:
C4H6O
InChi Key:
MLUCVPSAIODCQM-NSCUHMNNSA-N
SMILES:
C\C=C\C=O