CAS RN 1235560-28-7
CAS RN 1235560-28-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ ABT 639

CaV3.2 and CaV3.1 channel blocker (IC50 = 2.3 μM)
2
Medchem Express ABT-639, MedChemExpress
MedChemExpress ABT-639 is a novel, peripherally acting, selective T-type Ca2+ channel blocker.
