accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 124-64-1

ClOHPHOHOHO

CAS RN 124-64-1

IUPAC Name: tetrakis(hydroxymethyl)phosphanium chloride
Synonyms: tetrakis(hydroxymethyl)phosphonium chloride
Molecular Weight (g/mol): 190.56
Molecular Formula: C4H12ClO4P
InChi Key: AKXUUJCMWZFYMV-UHFFFAOYSA-M
SMILES: [Cl-].OC[P+](CO)(CO)CO
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products