CAS RN 1240257-17-3
CAS RN 1240257-17-3
IUPAC Name:
1-(bromomethyl)-2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms:
1-(bromomethyl)-2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol):
315.07
Molecular Formula:
C7H5BrF6S
InChi Key:
VZMCPUASCXQNFE-UHFFFAOYSA-N
SMILES:
FC1=C(CBr)C=CC(=C1)S(F)(F)(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Fluoro-4-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™
CAS: 1240257-17-3 Molecular Formula: C7H5BrF6S Molecular Weight (g/mol): 315.07 MDL Number: MFCD16652507 InChI Key: VZMCPUASCXQNFE-UHFFFAOYSA-N Synonym: 2-fluoro-4-pentafluorosulfur benzyl bromide,1-bromomethyl-2-fluoro-4-pentafluoro-??-sulfanyl benzene,2-fluoro-4-pentafluorothio benzyl bromide PubChem CID: 66523502 SMILES: FC1=C(CBr)C=CC(=C1)S(F)(F)(F)(F)F