accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1240257-25-3

SFFFFFFNH2

CAS RN 1240257-25-3

IUPAC Name: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 237.16
Molecular Formula: C6H5F6NS
InChi Key: VNFMUDAEGKTTOA-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products