accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1256482-73-1

CH3FSNH2

CAS RN 1256482-73-1

IUPAC Name: 4-fluoro-2-methylbenzene-1-carbothioamide
Synonyms: 4-fluoro-2-methylbenzenecarbothioamide
Molecular Weight (g/mol): 169.22
Molecular Formula: C8H8FNS
InChi Key: CIVOYVGIXSEIKD-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(F)=C1)C(N)=S

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products