accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 125867-25-6

CH3H3CCH3OHNNOO

CAS RN 125867-25-6

IUPAC Name: 2-(2,2-dimethylpropanamido)pyridine-3-carboxylate
Synonyms: 2-(2,2-dimethylpropanamido)pyridine-3-carboxylate
Molecular Weight (g/mol): 221.24
Molecular Formula: C11H13N2O3
InChi Key: KXLOGMIKCNUSNG-UHFFFAOYSA-M
SMILES: CC(C)(C)C(=O)NC1=NC=CC=C1C([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products