accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1259314-65-2

H3CNNNHNHClHCl

CAS RN 1259314-65-2

IUPAC Name: N-[4-(4-methylpiperazin-1-yl)phenyl]acridin-9-amine dihydrochloride
Synonyms: N-[4-(4-methylpiperazin-1-yl)phenyl]acridin-9-amine dihydrochloride
Molecular Weight (g/mol): 441.4
Molecular Formula: C24H26Cl2N4
InChi Key: VVZOADYFJAJZGL-UHFFFAOYSA-N
SMILES: Cl.Cl.CN1CCN(CC1)C1=CC=C(NC2=C3C=CC=CC3=NC3=CC=CC=C23)C=C1
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products