CAS RN 1260907-17-2
CAS RN 1260907-17-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ I-BET 762
Potent and high affinity BET bromodomain inhibitor; anti-inflammatory; orally bioavailable.
2
Medchem Express Molibresib, MedChemExpress
MedChemExpress Molibresib (I-BET762; GSK525762) is a BET bromodomain inhibitor with IC50 of 32.5-42.5 nM.
