accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 126318-27-2

OHONOO

CAS RN 126318-27-2

IUPAC Name: 3-nitro-1-benzofuran-5-ol
Synonyms: 3-nitro-1-benzofuran-5-ol
Molecular Weight (g/mol): 179.13
Molecular Formula: C8H5NO4
InChi Key: ZQPBTBWGCWHIGU-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC=C(C2=C1)[N+]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products