CAS RN 12672-29-6
CAS RN 12672-29-6
IUPAC Namn:
3,3',5,5'-tetrachloro-1,1'-biphenyl
Synonymer:
3,3',5,5'-tetrachlorobiphenyl 3,3',5,5'-tetrachlorodiphenyl
Molekylvikt (g/mol):
291.98
Molekylär sammansättning:
C12H6Cl4
InChi Key:
UTMWFJSRHLYRPY-UHFFFAOYSA-N
SMILES:
ClC1=CC(=CC(Cl)=C1)C1=CC(Cl)=CC(Cl)=C1
1
–
10
av
10
Resultat
8
Aroclor 1248 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
9
Aroclor 1248 100 μg/mL in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality