CAS RN 1276110-06-5
CAS RN 1276110-06-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express HS-173, MedChemExpress
MedChemExpress HS-173 is a novel PI3K inhibitor, that is used for cancer treatment.
