accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 128074-72-6

NONHNFOOHHCl

CAS RN 128074-72-6

IUPAC Name: 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate hydrochloride
Synonyms: ciprofloxacin hydrochloride ciprofloxacin monohydrochloride monohydrate ciprofloxacin hydrochloride hydrate ciprofloxacin hydrochloride monohydrate
Molecular Weight (g/mol): 385.82
Molecular Formula: C17H21ClFN3O4
InChi Key: ARPUHYJMCVWYCZ-UHFFFAOYSA-N
SMILES: O.Cl.OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products