CAS RN 129101-54-8
CAS RN 129101-54-8
IUPAC Name:
(2S,3S)-2,3-dihydroxybutanedioic acid; 3-[(1S)-1-(dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate
Synonyms:
D-tartaric acid; rivastigmine
Molecular Weight (g/mol):
400.43
Molecular Formula:
C18H28N2O8
InChi Key:
GWHQHAUAXRMMOT-RWALOXMOSA-N
SMILES:
O[C@@H]([C@H](O)C(O)=O)C(O)=O.CCN(C)C(=O)OC1=CC=CC(=C1)[C@H](C)N(C)C