CAS RN 129460-09-9
CAS RN 129460-09-9
IUPAC Name:
4-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Synonyms:
4-(tert-butoxy)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol):
411.45
Molecular Formula:
C23H25NO6
InChi Key:
VZXQYACYLGRQJU-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar N-Fmoc-L-aspartic acid 1-tert-butyl ester, 95%
CAS: 129460-09-9 Molecular Formula: C23H25NO6 Molecular Weight (g/mol): 411.45 MDL Number: MFCD00065631 InChI Key: VZXQYACYLGRQJU-UHFFFAOYNA-N Synonym: fmoc-asp-otbu,fmoc-asp-obut,s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid,l-fmoc-aspartic acid alpha-t-butyl ester,fmoc-asp oh-otbu,n-fmoc-l-aspartic acid 1-tert-butyl ester,1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate,3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid,1-tert-butyl n-fmoc-l-aspartate,fmoc asp-otbu PubChem CID: 7017910 SMILES: CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12