accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1302-42-7

NaOAlO

CAS RN 1302-42-7

IUPAC Name: sodium dioxoalumanuide
Synonyms: sodium aluminate
Molecular Weight (g/mol): 81.97
Molecular Formula: AlNaO2
InChi Key: IYJYQHRNMMNLRH-UHFFFAOYSA-N
SMILES: [Na+].O=[Al-]=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products