accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 130535-83-0

H3COHNOH

CAS RN 130535-83-0

IUPAC Name: N-(hydroxymethyl)nonanamide
Synonyms: N-(hydroxymethyl)nonanamide
Molecular Weight (g/mol): 187.28
Molecular Formula: C10H21NO2
InChi Key: NDENIKIKWXFRDN-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)NCO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products