CAS RN 13094-51-4
CAS RN 13094-51-4
IUPAC Name:
2-bromoacetyl 2-bromoacetate
Synonyms:
2-bromoacetyl 2-bromoacetate
Molecular Weight (g/mol):
259.88
Molecular Formula:
C4H4Br2O3
InChi Key:
FUKOTTQGWQVMQB-UHFFFAOYSA-N
SMILES:
BrCC(=O)OC(=O)CBr
1
–
1
of
1
results
1
Bromoacetic anhydride, 95%, Thermo Scientific Chemicals
CAS: 13094-51-4 Molecular Formula: C4H4Br2O3 Molecular Weight (g/mol): 259.87 MDL Number: MFCD00045041 InChI Key: FUKOTTQGWQVMQB-UHFFFAOYSA-N Synonym: bromoacetic anhydride,2-bromoacetic anhydride,acetic acid, 2-bromo-,1,1'-anhydride,2-bromoacetyl 2-bromoacetate,bromoaceticanhydride,acetic acid, bromo-, anhydride PubChem CID: 4259817 IUPAC Name: (2-bromoacetyl) 2-bromoacetate SMILES: C(C(=O)OC(=O)CBr)Br