accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1317-42-6

SCo

CAS RN 1317-42-6

IUPAC Name: λ²-cobalt(2+) sulfanediide
Synonyms: cobalt sulfide
Molecular Weight (g/mol): 90.99
Molecular Formula: CoS
InChi Key: INPLXZPZQSLHBR-UHFFFAOYSA-N
SMILES: [S--].[Co++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products