accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13322-19-5

CH3NNNHO

CAS RN 13322-19-5

IUPAC Name: (5-methyl-2-phenyl-2H-1,2,3-triazol-4-yl)methanol
Synonyms: (5-methyl-2-phenyl-1,2,3-triazol-4-yl)methanol
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: VYHJVMFMRIGUFV-UHFFFAOYSA-N
SMILES: CC1=NN(N=C1CO)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products