accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13362-78-2

NN(E)

CAS RN 13362-78-2

IUPAC Name: 4-[(1E)-2-(pyridin-4-yl)ethenyl]pyridine
Synonyms: 4-[(1E)-2-(pyridin-4-yl)ethenyl]pyridine
Molecular Weight (g/mol): 182.23
Molecular Formula: C12H10N2
InChi Key: MGFJDEHFNMWYBD-OWOJBTEDSA-N
SMILES: C(=C/C1=CC=NC=C1)\C1=CC=NC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products