CAS RN 133720-60-2
CAS RN 133720-60-2
IUPAC Name:
7-bromo-1-benzofuran
Synonyms:
7-bromo-1-benzofuran
Molecular Weight (g/mol):
197.03
Molecular Formula:
C8H5BrO
InChi Key:
GPZPQJITKRSVQJ-UHFFFAOYSA-N
SMILES:
BrC1=C2OC=CC2=CC=C1
1
–
2
of
2
results
2
7-Bromobenzo[b]furan, 97%, Thermo Scientific™
CAS: 133720-60-2 Molecular Formula: C8H5BrO Molecular Weight (g/mol): 197.03 MDL Number: MFCD09056756 InChI Key: GPZPQJITKRSVQJ-UHFFFAOYSA-N Synonym: 7-bromobenzofuran,7-bromobenzo b furan,benzofuran, 7-bromo,benzofuran, bromo,7-bromo-benzofuran,7-bromo-1-benzo b furan,acmc-1c18b PubChem CID: 113628 IUPAC Name: 7-bromo-1-benzofuran SMILES: BrC1=C2OC=CC2=CC=C1