accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 134296-40-5

HClCH3H3CNONOHNNH3C

CAS RN 134296-40-5

IUPAC Name: hydrogen N-{8-methyl-8-azabicyclo[3.2.1]octan-3-yl}-2-oxo-3-(propan-2-yl)-2,3-dihydro-1H-1,3-benzodiazole-1-carboxamide chloride
Synonyms: hydrogen 3-isopropyl-N-{8-methyl-8-azabicyclo[3.2.1]octan-3-yl}-2-oxo-1,3-benzodiazole-1-carboxamide chloride
Molecular Weight (g/mol): 378.9
Molecular Formula: C19H27ClN4O2
InChi Key: NQYXXIUVFVOJCX-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)N1C(=O)N(C(=O)NC2CC3CCC(C2)N3C)C2=CC=CC=C12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products