accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 134356-73-3

OF

CAS RN 134356-73-3

IUPAC Name: 2-(4-fluorophenyl)oxirane
Synonyms: 2-(4-fluorophenyl)oxirane
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: ICVNPQMUUHPPOK-UHFFFAOYNA-N
SMILES: FC1=CC=C(C=C1)C1CO1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products