accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 135065-71-3

CH3H3CCH3OONOOH

CAS RN 135065-71-3

IUPAC Name: tert-butyl 2-(hydroxymethyl)morpholine-4-carboxylate
Synonyms: tert-butyl 2-(hydroxymethyl)morpholine-4-carboxylate
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: FJYBLMJHXRWDAQ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCOC(CO)C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products