accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 135159-51-2

HClH3COONCH3H3COOOHO

CAS RN 135159-51-2

IUPAC Name: hydrogen 4-{[1-(dimethylamino)-3-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}propan-2-yl]oxy}-4-oxobutanoic acid chloride
Synonyms: hydrogen sarpogrelate chloride
Molecular Weight (g/mol): 465.97
Molecular Formula: C24H32ClNO6
InChi Key: POQBIDFFYCYHOB-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=CC=CC(CCC2=CC=CC=C2OCC(CN(C)C)OC(=O)CCC(O)=O)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products