accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13532-18-8

H3COOSCH3

CAS RN 13532-18-8

IUPAC Name: methyl 3-(methylsulfanyl)propanoate
Synonyms: methyl 3-(methylsulfanyl)propanoate
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: DMMJVMYCBULSIS-UHFFFAOYSA-N
SMILES: COC(=O)CCSC
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products