accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13593-03-8

CH3OPSOCH3ONN

CAS RN 13593-03-8

IUPAC Name: O,O-diethyl O-quinoxalin-2-yl phosphorothioate
Synonyms: savall sandoz 6538 spencer S-6538 chinalphos diethquinalphione diethquinalphion wie oben ekalux 25EC diethylquinalphione quinalphos 25 EC
Molecular Weight (g/mol): 298.3
Molecular Formula: C12H15N2O3PS
InChi Key: JYQUHIFYBATCCY-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CN=C2C=CC=CC2=N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products