CAS RN 1394820-69-9
CAS RN 1394820-69-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Ningetinib, MedChemExpress
MedChemExpress Ningetinib is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.
