accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 141109-19-5

H3COOClNHSHCl

CAS RN 141109-19-5

IUPAC Name: methyl 2-(2-chlorophenyl)-2-{[2-(thiophen-2-yl)ethyl]amino}acetate hydrochloride
Synonyms: methyl 2-(2-chlorophenyl)-2-{[2-(thiophen-2-yl)ethyl]amino}acetate hydrochloride
Molecular Weight (g/mol): 346.27
Molecular Formula: C15H17Cl2NO2S
InChi Key: ZXANKCFSGFEBQW-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(NCCC1=CC=CS1)C1=CC=CC=C1Cl
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products