accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14173-39-8

H2NClHOO

CAS RN 14173-39-8

IUPAC Name: 2-amino-3-(4-chlorophenyl)propanoic acid
Synonyms: fenclonine fenclonine hydrochloride fenclonine (L)- fenclonina P-chlorophenylalanine,DL 4-chloro-3-phenylalanine p-clorophenylalanine (D)-fenclonine fenclonine, (D)- fenchlonine
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: NIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(Cl)C=C1)C(O)=O
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products