CAS RN 142253-55-2
CAS RN 142253-55-2
IUPAC Name:
1-[(tert-butoxy)carbonyl]azetidine-3-carboxylate
Synonyms:
1-(tert-butoxycarbonyl)azetidine-3-carboxylate
Molecular Weight (g/mol):
200.22
Molecular Formula:
C9H14NO4
InChi Key:
NCADHSLPNSTDMJ-UHFFFAOYSA-M
SMILES:
CC(C)(C)OC(=O)N1CC(C1)C([O-])=O
1
–
2
of
2
results
1
1-Boc-azetidine-3-carboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 142253-55-2 Molecular Formula: C9H14NO4 Molecular Weight (g/mol): 200.22 MDL Number: MFCD01860897 InChI Key: NCADHSLPNSTDMJ-UHFFFAOYSA-M Synonym: 1-boc-azetidine-3-carboxylic acid,1-n-boc-3-azetidinecarboxylic acid,n-boc-azetidine-3-carboxylic acid,boc-azetidine-3-carboxylic acid,1-tert-butoxycarbonyl azetidine-3-carboxylic acid,1-tert-butoxy carbonyl azetidine-3-carboxylic acid,1-n-boc-azetidine-3-carboxylic acid,boc-aze 3-oh,azetidine-1,3-dicarboxylic acid mono-tert-butyl ester,n-boc-3-azetidine carboxylic acid PubChem CID: 2755981 SMILES: CC(C)(C)OC(=O)N1CC(C1)C([O-])=O