accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 142253-56-3

CH3H3CCH3OONOH

CAS RN 142253-56-3

IUPAC Name: tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: HXRDRJKAEYHOBB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(CO)C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products