CAS RN 1428630-85-6
CAS RN 1428630-85-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VU 0424465

Potent mGlu5 positive allosteric modulator and agonist; binds allosteric site with high affinity