CAS RN 1429624-84-9
CAS RN 1429624-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express PI4KIIIbeta-IN-9, MedChemExpress
MedChemExpress PI4KIIIbeta-IN-9 is a potent PI4KIIIβ inhibitor with an IC50 of 7 nM. PI4KIIIbeta-IN-9 also inhibits PI3Kδ and PI3Kγ with IC50s of 152 nM and 1046 nM, respectively.
