CAS RN 145-42-6
CAS RN 145-42-6
IUPAC Name:
sodium 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate
Synonyms:
sodium 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethanesulfonate
Molecular Weight (g/mol):
537.69
Molecular Formula:
C26H44NNaO7S
InChi Key:
JAJWGJBVLPIOOH-UHFFFAOYNA-M
SMILES:
[Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C
1
–
2
of
2
results
2
Taurocholic acid sodium salt, ≥97%, Practical grade, MP Biomedicals™
CAS: 145-42-6 Molecular Formula: C26H44NNaO7S Molecular Weight (g/mol): 537.688 InChI Key: JAJWGJBVLPIOOH-VXFFEJGCSA-M Synonym: sodium taurocholate,taurocholic acid sodium salt,taurocholate sodium,taurocholate sodium salt,unii-m6n3th81no,monosodium n-choloyltaurinate,m6n3th81no,monosodium taurocholate,sodium n-choloyltaurinate,monosodium taurocholic acid PubChem CID: 131632374 IUPAC Name: sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate SMILES: CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]