accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 145782-74-7

CH3SNNNH2NNOOPOOHOPOFFPOHOOHOHOHOHNaNaNaNa

CAS RN 145782-74-7

IUPAC Name: [({[({5-[6-amino-2-(propylsulfanyl)-9H-purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy)(hydroxy)phosphoryl]oxy}(hydroxy)phosphoryl)difluoromethyl]phosphonic acid tetrasodium
Synonyms: [({5-[6-amino-2-(propylsulfanyl)purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methoxy(hydroxy)phosphoryl)oxy(hydroxy)phosphoryl]difluoromethylphosphonic acid tetrasodium
Molecular Weight (g/mol): 707.29
Molecular Formula: C14H22F2N5Na4O12P3S
InChi Key: HNMHGAIGJBTXEA-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].[Na].CCCSC1=NC(N)=C2N=CN(C3OC(COP(O)(=O)OP(O)(=O)C(F)(F)P(O)(O)=O)C(O)C3O)C2=N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products