man

Test

CAS RN 14588-08-0

OPPPdOOOCH3CH3

CAS RN 14588-08-0

IUPAC Name: palladium(2+) bis(triphenylphosphane) diacetate
Synonyms: palladium(2+) bis(triphenylphosphine) diacetate
Molecular Weight (g/mol): 749.09
Molecular Formula: C40H36O4P2Pd
InChi Key: UVBXZOISXNZBLY-UHFFFAOYSA-L
SMILES: [Pd++].CC([O-])=O.CC([O-])=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results