accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1460-16-8

HOO

CAS RN 1460-16-8

IUPAC Name: cycloheptanecarboxylic acid
Synonyms: cycloheptanecarboxylic acid
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: VZFUCHSFHOYXIS-UHFFFAOYSA-N
SMILES: OC(=O)C1CCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products