accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14714-24-0

NNNOHOCl

CAS RN 14714-24-0

IUPAC Name: 6-chloroimidazo[1,2-b]pyridazine-2-carboxylic acid
Synonyms: 6-chloroimidazo[1,2-b]pyridazine-2-carboxylic acid
Molecular Weight (g/mol): 197.58
Molecular Formula: C7H4ClN3O2
InChi Key: HTUZJYQXKFDVLT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN2N=C(Cl)C=CC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products