CAS RN 1477949-42-0
CAS RN 1477949-42-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Santacruzamate A

Highly potent and selective HDAC2 inhibitor
2
Medchem Express Santacruzamate A, MedChemExpress
MedChemExpress Santacruzamate A (CAY-10683) is a potent and selective HDAC2 inhibitor with an IC50 of 119 pM.
