accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 148083-64-1

OONH(S)NHNOONOOOOH

CAS RN 148083-64-1

IUPAC Name: 6-[(2,4-dinitrophenyl)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 6-[(2,4-dinitrophenyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 534.53
Molecular Formula: C27H26N4O8
InChi Key: OENCMORJQAUAAJ-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products