accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1480833-70-2

CH3H3CNHNOFFFNH2

CAS RN 1480833-70-2

IUPAC Name: 2-{5-[4-(trifluoromethoxy)phenyl]-1H-imidazol-2-yl}propan-2-amine
Synonyms: 2-{4-[4-(trifluoromethoxy)phenyl]-3H-imidazol-2-yl}propan-2-amine
Molecular Weight (g/mol): 285.27
Molecular Formula: C13H14F3N3O
InChi Key: AGORGFNWYAUYSU-UHFFFAOYSA-N
SMILES: CC(C)(N)C1=NC=C(N1)C1=CC=C(OC(F)(F)F)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products