accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 148493-37-2

NIClCl

CAS RN 148493-37-2

IUPAC Name: 2,6-dichloro-3-iodopyridine
Synonyms: 2,6-dichloro-3-iodopyridine
Molecular Weight (g/mol): 273.88
Molecular Formula: C5H2Cl2IN
InChi Key: DPCQIHCGMIPSQV-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(I)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products