CAS RN 149923-10-4
CAS RN 149923-10-4
IUPAC Name:
(4-chlorophenyl)methanide; chlorozincylium
Synonyms:
(4-chlorophenyl)methanide; chlorozincylium
Molecular Weight (g/mol):
226.41
Molecular Formula:
C7H6Cl2Zn
InChi Key:
GIUOJMHOGPURPS-UHFFFAOYSA-M
SMILES:
Cl[Zn+].[CH2-]C1=CC=C(Cl)C=C1