man

Test

CAS RN 15201-61-3

OClOOOOClOOOOClOOOFe

CAS RN 15201-61-3

IUPAC Name: iron(3+) hydrate triperchlorate
Synonyms: iron(3+) hydrate triperchlorate
Molecular Weight (g/mol): 372.2
Molecular Formula: Cl3FeH2O13
InChi Key: PFPIMAZLJXJVAN-UHFFFAOYSA-K
SMILES: O.[Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results