accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15205-11-5

H2NFCl

CAS RN 15205-11-5

IUPAC Name: 1-(2-chloro-4-fluorophenyl)methanamine
Synonyms: 1-(2-chloro-4-fluorophenyl)methanamine
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: CBKWAXKMZUULLO-UHFFFAOYSA-N
SMILES: NCC1=CC=C(F)C=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products