accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15484-44-3

NaOSOOCl

CAS RN 15484-44-3

IUPAC Name: 2-chloroethane-1-sulfonic acid sodium
Synonyms: 2-chloroethanesulfonic acid sodium
Molecular Weight (g/mol): 167.56
Molecular Formula: C2H5ClNaO3S
InChi Key: SXGDKQFTZQEJMJ-UHFFFAOYSA-N
SMILES: [Na].OS(=O)(=O)CCCl
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products