accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1558-33-4

CH3SiClClCl

CAS RN 1558-33-4

IUPAC Name: dichloro(chloromethyl)methylsilane
Synonyms: dichloro(chloromethyl)methylsilane
Molecular Weight (g/mol): 163.5
Molecular Formula: C2H5Cl3Si
InChi Key: JAYBZWYBCUJLNQ-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products
1x1 image pixel for data collection